3-Fluoro-5-(pentafluorosulphur)benzonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-(pentafluorothio)benzonitrile

PC405670-1G
APOSPC405670-1GEA 348 EUR
APOSPC405670-1G
3-Fluoro-5-(pentafluorosulphur)benzonitrile 97%
3-Fluoro-5-(pentafluorosulphur)benzonitrile
Formel: C₇H₃F₆NS
Molecular Weight: 247,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201029
CAS-Nummer: 1373920-62-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR