2-Fluor-4-(pentafluorthio)benzaldehyd 97%

Lieferant: Apollo Scientific

PC405671-1G
APOSPC405671-1GEA 0 EUR
APOSPC405671-1G
2-Fluor-4-(pentafluorthio)benzaldehyd 97%
2-Fluoro-4-(pentafluorothio)benzaldehyde
Formel: C₇H₄F₆OS
Molecular Weight: 250,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652502
CAS-Nummer: 1800248-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR