2,6-Dichloro-4-(pentafluorosulphur)aniline 98%

Lieferant: Apollo Scientific
Achtung

PC405674-25G PC405674-5G PC405674-1G
APOSPC405674-25GEA 0 EUR
APOSPC405674-25G APOSPC405674-5G APOSPC405674-1G
2,6-Dichloro-4-(pentafluorosulphur)aniline 98%
2,6-Dichloro-4-(pentafluorosulphur)aniline
Formel: C₆H₄Cl₂F₅NS
Lagertemperatur: Kühlschrank
MDL: MFCD22201015
CAS-Nummer: 149757-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR