3-Chloro-5-(pentafluorosulphur)benzoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-(pentafluorothio)benzoic acid

PC405677-1G PC405677-5G
APOSPC405677-1GEA 422 EUR
APOSPC405677-1G APOSPC405677-5G
3-Chloro-5-(pentafluorosulphur)benzoic acid 97%
3-Chloro-5-(pentafluorosulphur)benzoic acid
Formel: C₇H₄ClF₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD16652415
CAS-Nummer: 1448317-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR