3-Fluor-5-(pentafluorthio)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC405678-1G
APOSPC405678-1GEA 841 EUR
APOSPC405678-1G
3-Fluor-5-(pentafluorthio)benzoesäure 98%
3-Fluoro-5-(pentafluorothio)benzoic acid
Formel: C₇H₄F₆O₂S
Molecular Weight: 266.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652421
CAS-Nummer: 1448319-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR