3-(Trifluoromethyl)bicyclo[1.1.1]pentane-1-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC405689-100MG
APOSPC405689-100MGEA 50.9 EUR
APOSPC405689-100MG
3-(Trifluoromethyl)bicyclo[1.1.1]pentane-1-carboxylic acid 97%
3-(Trifluoromethyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₇H₇F₃O₂
Molecular Weight: 180,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20544759
CAS-Nummer: 224584-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR