4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid

Lieferant: Apollo Scientific

PC405695-1G PC405695-500MG
APOSPC405695-1GEA 4080 EUR
APOSPC405695-1G APOSPC405695-500MG
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
Formel: C₁₂H₁₁FO₂
Molecular Weight: 206.213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037342
CAS-Nummer: 1980053-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR