4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
Lieferant: Apollo Scientific
PC405695-1G
PC405695-500MG
APOSPC405695-1GEA
4080
EUR
APOSPC405695-1G
APOSPC405695-500MG
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)benzoic acid
Formel:
C₁₂H₁₁FO₂ Molecular Weight: 206.213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29037342 CAS-Nummer: 1980053-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...