2-(Benzylamino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid

Lieferant: Apollo Scientific

PC405701-250MG
APOSPC405701-250MGEA 0 EUR
APOSPC405701-250MG
2-(Benzylamino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid
2-(Benzylamino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid
Formel: C₁₄H₁₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29037346
CAS-Nummer: 1980033-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR