2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid
Lieferant: Apollo Scientific
PC405704-250MG
APOSPC405704-250MGEA
1540
EUR
APOSPC405704-250MG
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid
Formel:
C₂₂H₂₀FNO₄ Molecular Weight: 381,397 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29037349 CAS-Nummer: 1980033-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...