4-Fluoropentylbenzene 99%

Lieferant: Apollo Scientific

PC4057E-1G PC4057E-5G
APOSPC4057E-1GEA 104 EUR
APOSPC4057E-1G APOSPC4057E-5G
4-Fluoropentylbenzene 99%
4-Fluoropentylbenzene
Formel: C₁₁H₁₅F
Molecular Weight: 166.24 g/mol
Siedepunkt: 105 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00079760
CAS-Nummer: 28593-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR