ω-Brom-4-fluoracetophenon 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-1-(4-fluorphenyl)ethan-1-on, Brommethyl-p-fluorphenylketon, 2-Brom-4'-fluoracetophenon, 4-Fluorphenacylbromid, α-Brom-4-fluoracetophenon
PC4060-25G
PC4060-5G
PC4060-100G
APOSPC4060-25GEA
43.5
EUR
APOSPC4060-25G
APOSPC4060-5G
APOSPC4060-100G
ω-Brom-4-fluoracetophenon 97%
ω-Brom-4-fluoracetophenon
Intermediate for preparation of 1,2-diarylcyclopentenolcyclooxygenase inhibitors: J.Med.Chem.,37,3878,1994.
Formel:
C₈H₆BrFO Molecular Weight: 217.04 g/mol Siedepunkt: 150 °C/12mmHg Schmelzpunkt: 45…49 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040830 CAS-Nummer: 403-29-2 EINECS: 206-955-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...