2-(2-Fluorphenyl)ethanol 99%
Lieferant: Apollo Scientific
Synonyme:
2-Fluorophenethyl alcohol
PC4066-25G
APOSPC4066-25GEA
154
EUR
APOSPC4066-25G
2-(2-Fluorphenyl)ethanol 99%
2-(2-Fluorphenyl)ethanol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol Dichte: 1,045 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002887 CAS-Nummer: 50919-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...