4-Hydroxy-2-n-propyl-6-trifluoromethylquinoline

Lieferant: Apollo Scientific

PC407024-1G
APOSPC407024-1GEA 393 EUR
APOSPC407024-1G
4-Hydroxy-2-n-propyl-6-trifluoromethylquinoline
4-Hydroxy-2-n-propyl-6-trifluoromethylquinoline
Formel: C₁₃H₁₂F₃NO
Molecular Weight: 255,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684130
CAS-Nummer: 927800-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR