(2-Nitro-5-trifluoromethylphenyl)acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-Nitro-5-(trifluoromethyl)phenylacetic acid

PC407026-1G
APOSPC407026-1GEA 256 EUR
APOSPC407026-1G
(2-Nitro-5-trifluoromethylphenyl)acetic acid
(2-Nitro-5-trifluoromethylphenyl)acetic acid
Formel: C₉H₆F₃NO₄
Molecular Weight: 249,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436986
CAS-Nummer: 927801-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR