1-Methyl-5-trifluoromethyl benzimidazole
Lieferant: Apollo Scientific
PC407055-1G
APOSPC407055-1GEA
285
EUR
APOSPC407055-1G
1-Methyl-5-trifluoromethyl benzimidazole
1-Methyl-5-trifluoromethyl benzimidazole
Formel:
C₉H₇F₃N₂ Molecular Weight: 200,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861887 CAS-Nummer: 53483-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...