5-Fluoro-1,2-dihydroisoquinolin-1-one ≥98%
Lieferant: Apollo Scientific
PC407077-250MG
APOSPC407077-250MGEA
95.7
EUR
APOSPC407077-250MG
5-Fluoro-1,2-dihydroisoquinolin-1-one ≥98%
5-Fluoro-1,2-dihydroisoquinolin-1-one
Formel:
C₉H₆FNO Lagertemperatur: Kühlschrank |
MDL:
MFCD12405865 CAS-Nummer: 410086-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...