3-Fluor-4-nitrophenol 97%

Lieferant: Apollo Scientific
Gefahr

PC4074-100G PC4074-25G PC4074-5G
APOSPC4074-100GEA 313 EUR
APOSPC4074-100G APOSPC4074-25G APOSPC4074-5G
3-Fluor-4-nitrophenol 97%
3-Fluor-4-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157.1 g/mol
Schmelzpunkt: 91…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041251
CAS-Nummer: 394-41-2
EINECS: 206-895-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR