3-Fluor-4-nitrophenol 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC4074-100G
PC4074-25G
PC4074-5G
APOSPC4074-100GEA
313
EUR
APOSPC4074-100G
APOSPC4074-25G
APOSPC4074-5G
3-Fluor-4-nitrophenol 97%
3-Fluor-4-nitrophenol
Formel:
C₆H₄FNO₃ Schmelzpunkt: 91…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041251 CAS-Nummer: 394-41-2 EINECS: 206-895-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...