2-(4-Fluorphenyl)thiophen 95%
Lieferant: Apollo Scientific
PC408001-100G
APOSPC408001-100GEA
74.1
EUR
APOSPC408001-100G
2-(4-Fluorphenyl)thiophen 95%
2-(4-Fluorophenyl)thiophene
Formel:
C₁₀H₇FS Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802535 CAS-Nummer: 58861-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...