(2-Chloro-6-fluoro-benzyl)-isopropyl-amine

Lieferant: Apollo Scientific

Synonyme: N-(2-Chloro-6-fluorobenzyl)propan-2-amine

PC408015-1G
APOSPC408015-1GEA 408 EUR
APOSPC408015-1G
(2-Chloro-6-fluoro-benzyl)-isopropyl-amine
(2-Chloro-6-fluoro-benzyl)-isopropyl-amine
Formel: C₁₀H₁₃ClFN
Molecular Weight: 201,668 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053432
CAS-Nummer: 62924-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR