1-(1-Chloro-ethyl)-4-fluoro-benzene

Lieferant: Apollo Scientific

Synonyme: 1-(1-Chloroethyl)-4-fluorobenzene

PC408016-1G
APOSPC408016-1GEA 423 EUR
APOSPC408016-1G
1-(1-Chloro-ethyl)-4-fluoro-benzene
1-(1-Chloro-ethyl)-4-fluoro-benzene
Formel: C₈H₈ClF
Molecular Weight: 158,601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06376752
CAS-Nummer: 456-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR