3-Fluoro-6-(methylamino)pyridine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Fluoro-N-methylpyridin-2-amine

PC408017-1G
APOSPC408017-1GEA 88.6 EUR
APOSPC408017-1G
3-Fluoro-6-(methylamino)pyridine
3-Fluoro-6-(methylamino)pyridine
Formel: C₆H₇FN₂
Molecular Weight: 126,132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848519
CAS-Nummer: 868636-72-0
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR