Cyclopropyl-(5-fluoro-pyrimidin-2-yl)-amine

Lieferant: Apollo Scientific

PC408018-1G
APOSPC408018-1GEA 202 EUR
APOSPC408018-1G
Cyclopropyl-(5-fluoro-pyrimidin-2-yl)-amine
Cyclopropyl-(5-fluoro-pyrimidin-2-yl)-amine
Formel: C₇H₈FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18837175
CAS-Nummer: 1289387-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR