N-Ethyl-3-fluorbenzylamin

Lieferant: Apollo Scientific

PC408019-1G
APOSPC408019-1GEA 0 EUR
APOSPC408019-1G
N-Ethyl-3-fluorbenzylamin
N-Ethyl-3-fluorbenzylamin
Formel: C₉H₁₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD06655514
CAS-Nummer: 90389-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR