2-Chloro-N-cyclopropyl-N-(2-fluoro-benzyl)-acetamide
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-N-cyclopropyl-N-(2-fluorobenzyl)acetamide
PC408020-500MG
APOSPC408020-500MGEA
865
EUR
APOSPC408020-500MG
2-Chloro-N-cyclopropyl-N-(2-fluoro-benzyl)-acetamide
2-Chloro-N-cyclopropyl-N-(2-fluoro-benzyl)-acetamide
Formel:
C₁₂H₁₃ClFNO Molecular Weight: 241,689 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07690515 CAS-Nummer: 874595-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...