{[Ethyl-(3-fluoro-benzyl)-carbamoyl]-methyl}-carbamic acid tert-butyl ester
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (2-(ethyl(3-fluorobenzyl)amino)-2-oxoethyl)carbamate
PC408021-500MG
APOSPC408021-500MGEA
945
EUR
APOSPC408021-500MG
{[Ethyl-(3-fluoro-benzyl)-carbamoyl]-methyl}-carbamic acid tert-butyl ester
{[Ethyl-(3-fluoro-benzyl)-carbamoyl]-methyl}-carbamic acid tert-butyl ester
Formel:
C₁₆H₂₃FN₂O₃ Molecular Weight: 310,364 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22631795 CAS-Nummer: 1417794-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...