Sodium (2-chloro-6-fluorophenyl)methanesulphinate

Lieferant: Apollo Scientific

PC408022-1G
APOSPC408022-1GEA 1080 EUR
APOSPC408022-1G
Sodium (2-chloro-6-fluorophenyl)methanesulphinate
Sodium (2-chloro-6-fluorophenyl)methanesulphinate
Formel: C₇H₅ClFO₂S·Na
Molecular Weight: 230.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631797
CAS-Nummer: 1417794-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR