5-Fluoro-N-methylpyridin-2-amine hydrochloride

Lieferant: Apollo Scientific

PC408023-1G
APOSPC408023-1GEA 873 EUR
APOSPC408023-1G
5-Fluoro-N-methylpyridin-2-amine hydrochloride
5-Fluoro-N-methylpyridin-2-amine hydrochloride
Formel: C₆H₇FN₂·ClH
Molecular Weight: 162,593 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631804
CAS-Nummer: 1417794-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR