Thioacetic acid S-(2-fluoro-benzyl) ester

Lieferant: Apollo Scientific

PC408024-1G
APOSPC408024-1GEA 823 EUR
APOSPC408024-1G
Thioacetic acid S-(2-fluoro-benzyl) ester
Thioacetic acid S-(2-fluoro-benzyl) ester
Formel: C₉H₉FOS
Lagertemperatur: Raumtemperatur
MDL: MFCD24369766
CAS-Nummer: 873463-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR