Thioacetic acid S-(3-fluoro-benzyl) ester

Lieferant: Apollo Scientific

Synonyme: S-3-Fluorobenzyl ethanethioate

PC408025-1G
APOSPC408025-1GEA 706 EUR
APOSPC408025-1G
Thioacetic acid S-(3-fluoro-benzyl) ester
Thioacetic acid S-(3-fluoro-benzyl) ester
Formel: C₉H₉FOS
Molecular Weight: 184,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369767
CAS-Nummer: 85582-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR