(5-Fluoro-pyridin-2-yl)-dimethyl-amine

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-N,N-dimethylpyridin-2-amine

PC408026-1G
APOSPC408026-1GEA 865 EUR
APOSPC408026-1G
(5-Fluoro-pyridin-2-yl)-dimethyl-amine
(5-Fluoro-pyridin-2-yl)-dimethyl-amine
Formel: C₇H₉FN₂
Molecular Weight: 140,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369772
CAS-Nummer: 1420961-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR