{(S)-1-[Cyclopropyl-(4-fluoro-benzyl)-carbamoyl]-ethyl}-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408027-500MG
APOSPC408027-500MGEA 1160 EUR
APOSPC408027-500MG
{(S)-1-[Cyclopropyl-(4-fluoro-benzyl)-carbamoyl]-ethyl}-carbamic acid tert-butyl ester
{(S)-1-[Cyclopropyl-(4-fluoro-benzyl)-carbamoyl]-ethyl}-carbamic acid tert-butyl ester
Formel: C₁₈H₂₅FN₂O₃
Molecular Weight: 336,401 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369776
CAS-Nummer: 1421010-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR