[Cyclopropyl-(4-fluoro-benzyl)-amino]-acetic acid methyl ester

Lieferant: Apollo Scientific

PC408028-1G
APOSPC408028-1GEA 1160 EUR
APOSPC408028-1G
[Cyclopropyl-(4-fluoro-benzyl)-amino]-acetic acid methyl ester
[Cyclopropyl-(4-fluoro-benzyl)-amino]-acetic acid methyl ester
Formel: C₁₃H₁₆FNO₂
Molecular Weight: 237,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13417328
CAS-Nummer: 1211709-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR