[Cyclopropyl-(4-fluoro-benzyl)-amino]acetic acid
Lieferant: Apollo Scientific
PC408029-500MG
APOSPC408029-500MGEA
655
EUR
APOSPC408029-500MG
[Cyclopropyl-(4-fluoro-benzyl)-amino]acetic acid
[Cyclopropyl-(4-fluoro-benzyl)-amino]acetic acid
Formel:
C₁₂H₁₄FNO₂ Molecular Weight: 223,243 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11937730 CAS-Nummer: 1181451-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...