2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol

Lieferant: Apollo Scientific

PC408030-500MG
APOSPC408030-500MGEA 386 EUR
APOSPC408030-500MG
2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol
2-[(4-Fluoro-benzyl)-methyl-amino]-ethanol
Formel: C₁₀H₁₄FNO
Molecular Weight: 183,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14646020
CAS-Nummer: 2248-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR