N-Cyclopropyl-1-(3-fluorophenyl)methanesulphonamide

Lieferant: Apollo Scientific

PC408032-1G
APOSPC408032-1GEA 556 EUR
APOSPC408032-1G
N-Cyclopropyl-1-(3-fluorophenyl)methanesulphonamide
N-Cyclopropyl-1-(3-fluorophenyl)methanesulphonamide
Formel: C₁₀H₁₂FNO₂S
Molecular Weight: 229,271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24382615
CAS-Nummer: 1420987-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR