5-Chlor-3-fluor-2-methoxypyridin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-5-chloro-3-fluoropyridine

PC408037-5G
APOSPC408037-5GEA 106 EUR
APOSPC408037-5G
5-Chlor-3-fluor-2-methoxypyridin 98%
5-Chlor-3-fluor-2-methoxypyridin
Formel: C₆H₅ClFNO
Molecular Weight: 161.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375056
CAS-Nummer: 886374-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR