2,2,3,3,4,4-Hexafluorobutyric acid
Lieferant: Apollo Scientific
|
Gefahr
|
PC408040-1G
APOSPC408040-1GEA
65.4
EUR
APOSPC408040-1G
2,2,3,3,4,4-Hexafluorobutyric acid
2,2,3,3,4,4-Hexafluorobutyric acid
Formel:
C₄H₂F₆O₂ Molecular Weight: 196,048 g/mol Siedepunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00465636 CAS-Nummer: 679-12-9 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...