2-Bromo-1,1-difluorocyclopropane

Achtung

PC408051-1G
APOSPC408051-1GEA 883 EUR
APOSPC408051-1G
2-Bromo-1,1-difluorocyclopropane
2-Bromo-1,1-difluorocyclopropane
Formel: C₃H₃BrF₂
Lagertemperatur: Kühlschrank
MDL: MFCD20622605
CAS-Nummer: 51326-64-8
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR