2,3-Difluoro-5-nitropyridine 98%

Lieferant: Apollo Scientific

PC408056-250MG PC408056-1G
APOSPC408056-250MGEA 98.9 EUR
APOSPC408056-250MG APOSPC408056-1G
2,3-Difluoro-5-nitropyridine 98%
2,3-Difluoro-5-nitropyridine
Formel: C₅H₂F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09750303
CAS-Nummer: 954219-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR