1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 96%
Lieferant: Apollo Scientific
PC408060-250MG
APOSPC408060-250MGEA
94.4
EUR
APOSPC408060-250MG
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 96%
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine
Formel:
C₅H₆F₃N₃ Molecular Weight: 165,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970058 CAS-Nummer: 1006436-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...