Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid

Lieferant: Apollo Scientific
Gefahr

PC408088-1G PC408088-5G
APOSPC408088-1GEA 109 EUR
APOSPC408088-1G APOSPC408088-5G
Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid
Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid
Formel: C₁₅HF₂₉O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD03412274
CAS-Nummer: 65150-95-0
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR