Potassium difluoroacetate 95%

Lieferant: Apollo Scientific
Achtung

PC408095-1G PC408095-5G
APOSPC408095-1GEA 147 EUR
APOSPC408095-1G APOSPC408095-5G
Potassium difluoroacetate 95%
Potassium difluoroacetate
Formel: C₂HF₂O₂·K
Lagertemperatur: Raumtemperatur
MDL: MFCD16620582
CAS-Nummer: 6291-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR