4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzonitrile
PC408124-5G
APOSPC408124-5GEA
302
EUR
APOSPC408124-5G
4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid
4-Cyan-2,3,5,6-tetrafluorbenzotrifluorid
Formel:
C₈F₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972776 CAS-Nummer: 15895-68-8 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...