1,4-Bis(bromomethyl)tetrafluorobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Bis(bromomethyl)-2,3,5,6-tetrafluorobenzene

PC408158-25G
APOSPC408158-25GEA 806 EUR
APOSPC408158-25G
1,4-Bis(bromomethyl)tetrafluorobenzene
1,4-Bis(bromomethyl)tetrafluorobenzene
Formel: C₈H₄Br₂F₄
Molecular Weight: 335.919 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07779350
CAS-Nummer: 776-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR