4-Bromo-1,1,1,2,2-pentafluorobutane

Lieferant: Apollo Scientific
Achtung

PC408202-5G
APOSPC408202-5GEA 321 EUR
APOSPC408202-5G
4-Bromo-1,1,1,2,2-pentafluorobutane
4-Bromo-1,1,1,2,2-pentafluorobutane
Formel: C₄H₄BrF₅
Molecular Weight: 226.971 g/mol
Siedepunkt: 76 °C
Lagertemperatur: Kühlschrank
MDL: MFCD16619618
CAS-Nummer: 52671-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR