4-Chloro-5,6-difluoronicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5,6-difluoropyridine-3-carboxylic acid

PC408223-1G
APOSPC408223-1GEA 524 EUR
APOSPC408223-1G
4-Chloro-5,6-difluoronicotinic acid 95%
4-Chloro-5,6-difluoronicotinic acid
Formel: C₆H₂ClF₂NO₂
Molecular Weight: 193,5400 g/mol
Schmelzpunkt: 117…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09031172
CAS-Nummer: 851386-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR