2-Fluoro-5-hydroxyphenylacetic acid

Lieferant: Apollo Scientific
Achtung

PC408247-1G
APOSPC408247-1GEA 232 EUR
APOSPC408247-1G
2-Fluoro-5-hydroxyphenylacetic acid
2-Fluoro-5-hydroxyphenylacetic acid
Formel: C₈H₇FO₃
Molecular Weight: 170,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263179
CAS-Nummer: 149029-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR