2-Fluoro-3-methoxybenzylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-Fluoro-3-methoxyphenyl)methylamine

PC408258-1G PC408258-5G
APOSPC408258-1GEA 108 EUR
APOSPC408258-1G APOSPC408258-5G
2-Fluoro-3-methoxybenzylamine
2-Fluoro-3-methoxybenzylamine
Formel: C₈H₁₀FNO
Molecular Weight: 155.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11045854
CAS-Nummer: 93071-81-9
UN: 3267
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR